2-Fluoro-4-methyl-5-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303389-1G
APOSPC303389-1GEA 474 EUR
APOSPC303389-1G
2-Fluoro-4-methyl-5-(trifluoromethyl)benzoic acid 97%
2-Fluoro-4-methyl-5-(trifluoromethyl)benzoic acid
Formel: C₉H₆F₄O₂
Molecular Weight: 222,136 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD31916397
CAS-Nummer: 2385359-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR