4-Chloro-3-(trifluoromethylthio)phenylacetonitrile 97%

Lieferant: Apollo Scientific

PC303392-1G
APOSPC303392-1GEA 0 EUR
APOSPC303392-1G
4-Chloro-3-(trifluoromethylthio)phenylacetonitrile 97%
4-Chloro-3-(trifluoromethylthio)phenylacetonitrile
Formel: C₉H₅ClF₃NS
Lagertemperatur: Raumtemperatur
MDL: MFCD32640053
CAS-Nummer: 2514625-02-4
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR