4-Chloro-3-(trifluoromethylthio)phenylacetic acid

Lieferant: Apollo Scientific

PC303393-1G
APOSPC303393-1GEA 348 EUR
APOSPC303393-1G
4-Chloro-3-(trifluoromethylthio)phenylacetic acid
4-Chloro-3-(trifluoromethylthio)phenylacetic acid
Formel: C₉H₆ClF₃O₂S
Molecular Weight: 270,656 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2415751-73-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR