4-Chloro-3-(trifluoromethylthio)benzylamine 98%

Lieferant: Apollo Scientific

PC303394-1G
APOSPC303394-1GEA 290 EUR
APOSPC303394-1G
4-Chloro-3-(trifluoromethylthio)benzylamine 98%
4-Chloro-3-(trifluoromethylthio)benzylamine
Formel: C₈H₇ClF₃NS
Molecular Weight: 241,661 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640055
CAS-Nummer: 2415751-71-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR