3-Fluoro-2,4-dimethylbenzylamine 98%

Lieferant: Apollo Scientific

PC303398-1G
APOSPC303398-1GEA 290 EUR
APOSPC303398-1G
3-Fluoro-2,4-dimethylbenzylamine 98%
3-Fluoro-2,4-dimethylbenzylamine
Formel: C₉H₁₂FN
Molecular Weight: 153,197 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23712164
CAS-Nummer: 1427366-15-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR