3-Fluoro-2,4-dimethylbenzaldehyde 98%

Lieferant: Apollo Scientific

Synonyme: 2,4-Dimethyl-3-fluorobenzaldehyde

PC303401-1G
APOSPC303401-1GEA 144 EUR
APOSPC303401-1G
3-Fluoro-2,4-dimethylbenzaldehyde 98%
3-Fluoro-2,4-dimethylbenzaldehyde
Formel: C₉H₉FO
Molecular Weight: 152,166 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04972838
CAS-Nummer: 363134-36-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR