2-Methoxy-4-(trifluoromethoxy)cinnamic acid 97%

Lieferant: Apollo Scientific

PC303402-1G
APOSPC303402-1GEA 145 EUR
APOSPC303402-1G
2-Methoxy-4-(trifluoromethoxy)cinnamic acid 97%
2-Methoxy-4-(trifluoromethoxy)cinnamic acid
Formel: C₁₁H₉F₃O₄
Molecular Weight: 262,182 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18398498
CAS-Nummer: 1262019-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR