6-Chloro-2,4-difluoro-3-methylbenzamide 97%

Lieferant: Apollo Scientific

PC303405-1G PC303405-250MG
APOSPC303405-1GEA 927 EUR
APOSPC303405-1G APOSPC303405-250MG
6-Chloro-2,4-difluoro-3-methylbenzamide 97%
6-Chloro-2,4-difluoro-3-methylbenzamide
Formel: C₈H₆ClF₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD32640059
CAS-Nummer: 2415751-80-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR