6-Chloro-2,4-difluoro-3-methylbenzonitrile 97%

Lieferant: Apollo Scientific

PC303406-1G PC303406-250MG
APOSPC303406-1GEA 975 EUR
APOSPC303406-1G APOSPC303406-250MG
6-Chloro-2,4-difluoro-3-methylbenzonitrile 97%
6-Chloro-2,4-difluoro-3-methylbenzonitrile
Formel: C₈H₄ClF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD32640060
CAS-Nummer: 2415751-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR