4-Chloro-3-fluoro-2-methyl-6-(trifluoromethyl)aniline 97%

Lieferant: Apollo Scientific

PC303407-1G
APOSPC303407-1GEA 377 EUR
APOSPC303407-1G
4-Chloro-3-fluoro-2-methyl-6-(trifluoromethyl)aniline 97%
4-Chloro-3-fluoro-2-methyl-6-(trifluoromethyl)aniline
Formel: C₈H₆ClF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD32640061
CAS-Nummer: 2415751-74-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR