6-Chloro-2,4-difluoro-3-methylaniline 97%

Lieferant: Apollo Scientific

PC303420-1G PC303420-250MG
APOSPC303420-1GEA 1050 EUR
APOSPC303420-1G APOSPC303420-250MG
6-Chloro-2,4-difluoro-3-methylaniline 97%
6-Chloro-2,4-difluoro-3-methylaniline
Formel: C₇H₆ClF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD32640069
CAS-Nummer: 2300095-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR