2,5-Difluoro-4-methoxyaniline 97%
Lieferant: Apollo Scientific
PC303423-1G
APOSPC303423-1GEA
368
EUR
APOSPC303423-1G
2,5-Difluoro-4-methoxyaniline 97%
2,5-Difluoro-4-methoxyaniline
Formel:
C₇H₇F₂NO Molecular Weight: 159,133 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18394891 CAS-Nummer: 394223-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...