5-Bromo-2,3-difluorophenylacetonitrile 97%

Lieferant: Apollo Scientific

PC303425-1G
APOSPC303425-1GEA 290 EUR
APOSPC303425-1G
5-Bromo-2,3-difluorophenylacetonitrile 97%
5-Bromo-2,3-difluorophenylacetonitrile
Formel: C₈H₄BrF₂N
Molecular Weight: 232,025 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28738339
CAS-Nummer: 1780864-29-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR