5-Bromo-2,3-difluorocinnamic acid

Lieferant: Apollo Scientific

Synonyme: (E)-3-(5-bromo-2,3-difluorophenyl)acrylic acid

PC303428-1G PC303428-5G
APOSPC303428-1GEA 209 EUR
APOSPC303428-1G APOSPC303428-5G
5-Bromo-2,3-difluorocinnamic acid
5-Bromo-2,3-difluorocinnamic acid
Formel: C₉H₅BrF₂O₂
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2387460-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR