5-Bromo-2,3-difluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303431-1G
APOSPC303431-1GEA 812 EUR
APOSPC303431-1G
5-Bromo-2,3-difluorobenzenesulphonyl chloride
5-Bromo-2,3-difluorobenzenesulphonyl chloride
Formel: C₆H₂BrClF₂O₂S
Molecular Weight: 291,498 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD28737881
CAS-Nummer: 1805119-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR