6-Bromo-2,3-difluorophenylacetonitrile 97%

Lieferant: Apollo Scientific

Synonyme: 2-(6-Bromo-2,3-difluorophenyl)acetonitrile

PC303433-1G PC303433-5G
APOSPC303433-1GEA 145 EUR
APOSPC303433-1G APOSPC303433-5G
6-Bromo-2,3-difluorophenylacetonitrile 97%
6-Bromo-2,3-difluorophenylacetonitrile
Formel: C₈H₄BrF₂N
Lagertemperatur: Raumtemperatur
MDL: MFCD26686146
CAS-Nummer: 1517576-71-4
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR