3-Bromo-2,6-difluorobenzamide 97%

Lieferant: Apollo Scientific

PC303451-1G
APOSPC303451-1GEA 279 EUR
APOSPC303451-1G
3-Bromo-2,6-difluorobenzamide 97%
3-Bromo-2,6-difluorobenzamide
Formel: C₇H₄BrF₂NO
Molecular Weight: 236,014 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26680221
CAS-Nummer: 1541810-83-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR