5-Bromo-3-chloro-2-fluorobenzoyl chloride

Lieferant: Apollo Scientific

PC303458-1G
APOSPC303458-1GEA 0 EUR
APOSPC303458-1G
5-Bromo-3-chloro-2-fluorobenzoyl chloride
5-Bromo-3-chloro-2-fluorobenzoyl chloride
Formel: C₇H₂BrCl₂FO
Lagertemperatur: Raumtemperatur
MDL: MFCD26686459
CAS-Nummer: 1536225-35-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR