5-Bromo-3-chloro-2-fluorobenzamide

Lieferant: Apollo Scientific

PC303459-1G
APOSPC303459-1GEA 654 EUR
APOSPC303459-1G
5-Bromo-3-chloro-2-fluorobenzamide
5-Bromo-3-chloro-2-fluorobenzamide
Formel: C₇H₄BrClFNO
Molecular Weight: 252,468 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640077
CAS-Nummer: 1521360-71-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR