5-Bromo-3-chloro-2-fluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303462-1G
APOSPC303462-1GEA 1050 EUR
APOSPC303462-1G
5-Bromo-3-chloro-2-fluorobenzenesulphonyl chloride
5-Bromo-3-chloro-2-fluorobenzenesulphonyl chloride
Formel: C₆H₂BrCl₂FO₂S
Molecular Weight: 307,952 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640078
CAS-Nummer: 2386664-25-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR