3-Bromo-2-fluoro-5-(trifluoromethyl)benzylamine

Lieferant: Apollo Scientific

PC303469-1G PC303469-5G
APOSPC303469-1GEA 418 EUR
APOSPC303469-1G APOSPC303469-5G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzylamine
3-Bromo-2-fluoro-5-(trifluoromethyl)benzylamine
Formel: C₈H₆BrF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD20485012
CAS-Nummer: 871325-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR