3-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303470-1G PC303470-5G
APOSPC303470-1GEA 0 EUR
APOSPC303470-1G APOSPC303470-5G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzoic acid
Formel: C₈H₃BrF₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD31382551
CAS-Nummer: 2091606-48-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR