3-Bromo-2-fluoro-5-(trifluoromethyl)benzamide 97%

Lieferant: Apollo Scientific

PC303472-1G
APOSPC303472-1GEA 279 EUR
APOSPC303472-1G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzamide 97%
3-Bromo-2-fluoro-5-(trifluoromethyl)benzamide
Formel: C₈H₄BrF₄NO
Lagertemperatur: Raumtemperatur
MDL: MFCD32265222
CAS-Nummer: 2092800-03-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR