3-Bromo-2-fluoro-5-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303475-1G
APOSPC303475-1GEA 1050 EUR
APOSPC303475-1G
3-Bromo-2-fluoro-5-(trifluoromethyl)benzenesulphonyl chloride
3-Bromo-2-fluoro-5-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₂BrClF₄O₂S
Molecular Weight: 341,505 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640080
CAS-Nummer: 2387196-53-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR