5-Bromo-4-chloro-2-fluorobenzyl bromide 97%

Lieferant: Apollo Scientific

PC303478-10G PC303478-1G PC303478-5G
APOSPC303478-10GEA 1350 EUR
APOSPC303478-10G APOSPC303478-1G APOSPC303478-5G
5-Bromo-4-chloro-2-fluorobenzyl bromide 97%
5-Bromo-4-chloro-2-fluorobenzyl bromide
Formel: C₇H₄Br₂ClF
Lagertemperatur: Raumtemperatur
MDL: MFCD28060816
CAS-Nummer: 1823871-15-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR