5-Bromo-4-chloro-2-fluoroaniline

Lieferant: Apollo Scientific

PC303485-1G PC303485-5G PC303485-250MG
APOSPC303485-1GEA 0 EUR
APOSPC303485-1G APOSPC303485-5G APOSPC303485-250MG
5-Bromo-4-chloro-2-fluoroaniline
5-Bromo-4-chloro-2-fluoroaniline
Formel: C₆H₄BrClFN
Lagertemperatur: Raumtemperatur
MDL: MFCD00274773
CAS-Nummer: 111010-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR