5-Bromo-4-chloro-2-fluorobenzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303486-1G
APOSPC303486-1GEA 0 EUR
APOSPC303486-1G
5-Bromo-4-chloro-2-fluorobenzenesulphonyl chloride
5-Bromo-4-chloro-2-fluorobenzenesulphonyl chloride
Formel: C₆H₂BrCl₂FO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD19441846
CAS-Nummer: 1070972-67-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR