3-Bromo-5,6-difluoro-2-methoxybenzaldehyde 97%
Lieferant: Apollo Scientific
PC303487-1G
PC303487-5G
PC303487-25G
PC303487-100G
APOSPC303487-1GEA
198
EUR
APOSPC303487-1G
APOSPC303487-5G
APOSPC303487-25G
APOSPC303487-100G
3-Bromo-5,6-difluoro-2-methoxybenzaldehyde 97%
3-Bromo-5,6-difluoro-2-methoxybenzaldehyde
Formel:
C₈H₅BrF₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD28629022 CAS-Nummer: 1782614-60-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...