3-Bromo-5,6-difluoro-2-methoxybenzaldehyde 97%

Lieferant: Apollo Scientific

PC303487-1G PC303487-5G PC303487-25G PC303487-100G
APOSPC303487-1GEA 198 EUR
APOSPC303487-1G APOSPC303487-5G APOSPC303487-25G APOSPC303487-100G
3-Bromo-5,6-difluoro-2-methoxybenzaldehyde 97%
3-Bromo-5,6-difluoro-2-methoxybenzaldehyde
Formel: C₈H₅BrF₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD28629022
CAS-Nummer: 1782614-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR