3-Bromo-5,6-difluoro-2-methoxyphenylacetonitrile

Lieferant: Apollo Scientific

PC303490-1G PC303490-5G
APOSPC303490-1GEA 325 EUR
APOSPC303490-1G APOSPC303490-5G
3-Bromo-5,6-difluoro-2-methoxyphenylacetonitrile
3-Bromo-5,6-difluoro-2-methoxyphenylacetonitrile
Formel: C₉H₆BrF₂NO
Lagertemperatur: Raumtemperatur
MDL: MFCD32640084
CAS-Nummer: 1892087-78-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR