3-Bromo-5-chloro-2,6-difluorobenzaldehyde 97%
Lieferant: Apollo Scientific
PC303514-1G
APOSPC303514-1GEA
198
EUR
APOSPC303514-1G
3-Bromo-5-chloro-2,6-difluorobenzaldehyde 97%
3-Bromo-5-chloro-2,6-difluorobenzaldehyde
Formel:
C₇H₂BrClF₂O Molecular Weight: 255,444 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12545924 CAS-Nummer: 2514963-78-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...