3-Bromo-5-chloro-2,6-difluorophenylacetonitrile 97%

Lieferant: Apollo Scientific

PC303517-1G
APOSPC303517-1GEA 325 EUR
APOSPC303517-1G
3-Bromo-5-chloro-2,6-difluorophenylacetonitrile 97%
3-Bromo-5-chloro-2,6-difluorophenylacetonitrile
Formel: C₈H₃BrClF₂N
Molecular Weight: 266,47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640107
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR