3-Bromo-4-(trifluoromethoxy)benzylamine 98%

Lieferant: Apollo Scientific

PC303527-1G PC303527-5G
APOSPC303527-1GEA 0 EUR
APOSPC303527-1G APOSPC303527-5G
3-Bromo-4-(trifluoromethoxy)benzylamine 98%
3-Bromo-4-(trifluoromethoxy)benzylamine
Formel: C₈H₇BrF₃NO
Lagertemperatur: Raumtemperatur
MDL: MFCD31720102
CAS-Nummer: 1699274-98-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR