5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide 97%

Lieferant: Apollo Scientific

PC303542-1G PC303542-5G
APOSPC303542-1GEA 0 EUR
APOSPC303542-1G APOSPC303542-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide 97%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzyl bromide
Formel: C₈H₄Br₂F₄
Lagertemperatur: Raumtemperatur
MDL: MFCD32640123
CAS-Nummer: 2090283-06-8
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR