5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetonitrile

Lieferant: Apollo Scientific

PC303543-1G PC303543-5G
APOSPC303543-1GEA 0 EUR
APOSPC303543-1G APOSPC303543-5G
5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetonitrile
5-Bromo-2-fluoro-3-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₄BrF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD32640124
CAS-Nummer: 2092340-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR