5-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid 97%

Lieferant: Apollo Scientific

PC303546-1G PC303546-5G PC303546-25G
APOSPC303546-1GEA 0 EUR
APOSPC303546-1G APOSPC303546-5G APOSPC303546-25G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid 97%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzoic acid
Formel: C₈H₃BrF₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD29043332
CAS-Nummer: 1881295-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR