5-Bromo-2-fluoro-3-(trifluoromethyl)benzamide 97%
Lieferant: Apollo Scientific
PC303548-1G
APOSPC303548-1GEA
290
EUR
APOSPC303548-1G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzamide 97%
5-Bromo-2-fluoro-3-(trifluoromethyl)benzamide
Formel:
C₈H₄BrF₄NO Molecular Weight: 286,021 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640128 CAS-Nummer: 2090465-22-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...