5-Bromo-2-fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
Lieferant: Apollo Scientific
PC303550-1G
APOSPC303550-1GEA
1060
EUR
APOSPC303550-1G
5-Bromo-2-fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
5-Bromo-2-fluoro-3-(trifluoromethyl)benzenesulphonyl chloride
Formel:
C₇H₂BrClF₄O₂S Molecular Weight: 341,505 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640129 CAS-Nummer: 2384155-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...