2-Bromo-5-fluoro-4-(trifluoromethyl)benzaldehyde 97%

Lieferant: Apollo Scientific

PC303552-1G PC303552-5G
APOSPC303552-1GEA 209 EUR
APOSPC303552-1G APOSPC303552-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzaldehyde 97%
2-Bromo-5-fluoro-4-(trifluoromethyl)benzaldehyde
Formel: C₈H₃BrF₄O
Lagertemperatur: Raumtemperatur
MDL: MFCD32640130
CAS-Nummer: 2091605-30-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR