2-Bromo-5-fluoro-4-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303561-1G
APOSPC303561-1GEA 812 EUR
APOSPC303561-1G
2-Bromo-5-fluoro-4-(trifluoromethyl)benzenesulphonyl chloride
2-Bromo-5-fluoro-4-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₂BrClF₄O₂S
Molecular Weight: 341,505 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640139
CAS-Nummer: 2383916-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR