2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetonitrile

Lieferant: Apollo Scientific

PC303562-1G PC303562-5G
APOSPC303562-1GEA 0 EUR
APOSPC303562-1G APOSPC303562-5G
2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetonitrile
2-Bromo-5-fluoro-4-(trifluoromethyl)phenylacetonitrile
Formel: C₉H₄BrF₄N
Lagertemperatur: Raumtemperatur
MDL: MFCD32640140
CAS-Nummer: 2091737-50-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR