3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide

Lieferant: Apollo Scientific

PC303565-1G PC303565-5G
APOSPC303565-1GEA 0 EUR
APOSPC303565-1G APOSPC303565-5G
3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide
3-Bromo-4-chloro-5-(trifluoromethyl)benzyl bromide
Formel: C₈H₄Br₂ClF₃
Lagertemperatur: Raumtemperatur
MDL: MFCD32640143
CAS-Nummer: 2386636-01-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR