3-Bromo-4-chloro-5-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303574-1G
APOSPC303574-1GEA 754 EUR
APOSPC303574-1G
3-Bromo-4-chloro-5-(trifluoromethyl)benzenesulphonyl chloride
3-Bromo-4-chloro-5-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₂BrCl₂F₃O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD32640151
CAS-Nummer: 2386905-65-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR