3-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol

Lieferant: Apollo Scientific

PC303576-1G PC303576-5G
APOSPC303576-1GEA 231 EUR
APOSPC303576-1G APOSPC303576-5G
3-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol
3-Bromo-2-chloro-5-(trifluoromethyl)benzyl alcohol
Formel: C₈H₅BrClF₃O
Molecular Weight: 289.477 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640153
CAS-Nummer: 2092609-70-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR