3-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid 97%
Lieferant: Apollo Scientific
PC303581-1G
APOSPC303581-1GEA
198
EUR
APOSPC303581-1G
3-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid 97%
3-Bromo-2-chloro-5-(trifluoromethyl)benzoic acid
Formel:
C₈H₃BrClF₃O₂ Molecular Weight: 303,46 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640158 CAS-Nummer: 2091224-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...