3-Bromo-2-chloro-5-(trifluoromethyl)benzonitrile 97%

Lieferant: Apollo Scientific

PC303584-1G
APOSPC303584-1GEA 325 EUR
APOSPC303584-1G
3-Bromo-2-chloro-5-(trifluoromethyl)benzonitrile 97%
3-Bromo-2-chloro-5-(trifluoromethyl)benzonitrile
Formel: C₈H₂BrClF₃N
Molecular Weight: 284,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640161
CAS-Nummer: 133013-31-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR