5-Bromo-2-methyl-3-(trifluoromethyl)benzoic acid 97%
Lieferant: Apollo Scientific
PC303592-1G
PC303592-5G
APOSPC303592-1GEA
0
EUR
APOSPC303592-1G
APOSPC303592-5G
5-Bromo-2-methyl-3-(trifluoromethyl)benzoic acid 97%
5-Bromo-2-methyl-3-(trifluoromethyl)benzoic acid
Formel:
C₉H₆BrF₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD27942784 CAS-Nummer: 1266728-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...