5-Bromo-2-methyl-3-(trifluoromethyl)benzamide 97%

Lieferant: Apollo Scientific

PC303594-1G
APOSPC303594-1GEA 348 EUR
APOSPC303594-1G
5-Bromo-2-methyl-3-(trifluoromethyl)benzamide 97%
5-Bromo-2-methyl-3-(trifluoromethyl)benzamide
Formel: C₉H₇BrF₃NO
Molecular Weight: 282,057 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640168
CAS-Nummer: 2387008-53-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR