5-Bromo-2-methyl-3-(trifluoromethyl)benzenesulphonyl chloride
Lieferant: Apollo Scientific
PC303597-1G
APOSPC303597-1GEA
0
EUR
APOSPC303597-1G
5-Bromo-2-methyl-3-(trifluoromethyl)benzenesulphonyl chloride
5-Bromo-2-methyl-3-(trifluoromethyl)benzenesulphonyl chloride
Formel:
C₈H₅BrClF₃O₂S Lagertemperatur: Raumtemperatur |
MDL:
MFCD32640170 CAS-Nummer: 2385598-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...