3-Bromo-2-chloro-5-(trifluoromethyl)benzenesulphonyl chloride

Lieferant: Apollo Scientific

PC303598-1G
APOSPC303598-1GEA 754 EUR
APOSPC303598-1G
3-Bromo-2-chloro-5-(trifluoromethyl)benzenesulphonyl chloride
3-Bromo-2-chloro-5-(trifluoromethyl)benzenesulphonyl chloride
Formel: C₇H₂BrCl₂F₃O₂S
Molecular Weight: 357.96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD32640171
CAS-Nummer: 2384426-01-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR